missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole-d4, TRC

Product Code. 30406432
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406432 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30406432 Supplier Toronto Research Chemicals Supplier No. TRCF4210021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluconazole-D4
CAS 1124197-58-5
Purity Grade Notes HPLC
Molecular Formula C13 2H4 H8 F2 N6 O
Quantity 1 mg
Synonym Fluconazole D4 (bismethylene D4), 1H-1,2,4-Triazole-1-ethan-β,β-d2-ol, α-(2,4-difluorophenyl)-α-(1H-1,2,4-triazol-1-ylmethyl-d2)- (ACI), α-(2,4-Difluorophenyl)-α-(1H-1,2,4-triazol-1-ylmethyl-d2)-1H-1,2,4-triazole-1-ethan-β,β-d2-ol (ACI)
InChI Formula InChI=1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2/i4D2,5D2
SMILES [2H]C([2H])(n1cncn1)C(O)(c2ccc(F)cc2F)C([2H])([2H])n3cncn3
IUPAC Name 1,1,3,3-tetradeuterio-2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
Molecular Weight (g/mol) 310.3
Formula Weight 310.1292
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.