missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluconazole-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Fluconazole-D4 |
| CAS | 1124197-58-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 2H4 H8 F2 N6 O |
| Quantity | 10 mg |
| Synonym | Fluconazole D4 (bismethylene D4), 1H-1,2,4-Triazole-1-ethan-β,β-d2-ol, α-(2,4-difluorophenyl)-α-(1H-1,2,4-triazol-1-ylmethyl-d2)- (ACI), α-(2,4-Difluorophenyl)-α-(1H-1,2,4-triazol-1-ylmethyl-d2)-1H-1,2,4-triazole-1-ethan-β,β-d2-ol (ACI) |
| InChI Formula | InChI=1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2/i4D2,5D2 |
| SMILES | [2H]C([2H])(n1cncn1)C(O)(c2ccc(F)cc2F)C([2H])([2H])n3cncn3 |
| IUPAC Name | 1,1,3,3-tetradeuterio-2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol |
| Molecular Weight (g/mol) | 310.3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?