missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole-d4 (bismethylene-d4), CDN

Product Code. 30317545
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Product Code. Quantity unitSize
30317545 0.005 g 0.01g
30311792 0.01 g 0.01g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30317545

Brand: CDN CDND61430.005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluconazole-D4
CAS 1124197-58-5
Molecular Formula C13 2H4 H8 F2 N6 O
Quantity 0.005 g
InChI Formula InChI=1 S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22 H,4-5H2/i4D2,5D2
SMILES [2 H]C([2 H])(n1cncn1)C(O)(c2ccc(F)cc2F)C([2 H])([2 H])n3cncn3
IUPAC Name 1,1,3,3-tetradeuterio-2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
Molecular Weight (g/mol) 310.3
Formula Weight 310.1292 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.