missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole, TRC

Product Code. 30011287
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011287 1 g 1g
17930345 100 mg 100mg
17922805 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011287 Supplier Toronto Research Chemicals Supplier No. TRCF4185001G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31430-15-6
Molecular Formula C16 H12 F N3 O3
Molecular Weight (g/mol) 313.28
Synonym Flubendazole, N-[6-(4-Fluorobenzoyl)-1H-benzimidazol-2-yl]carbamic acid methyl ester, 5-(p-Fluorobenzoyl)-2-benzimidazolecarbamic acid methyl ester, [5-(4-Fluorobenzoyl)-1H-benzimidazol-2-yl]carbamic acid methyl ester, Flubenol, Flubenvet, Flumoxal, Flumoxane, Fluvermal, Methyl (5-p-fluorobenzoyl-2-benzimidazolyl)carbamate, NSC 313680, R 17889, R 17899
IUPAC Name methyl N-[5-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(F)cc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H12FN3O3/c1-23-16(22)20-15-18-12-7-4-10(8-13(12)19-15)14(21)9-2-5-11(17)6-3-9/h2-8H,1H3,(H2,18,19,20,22)
Formula Weight 313.0863
Percent Purity >95%
Chemical Name or Material Flubendazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.