missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole, MedChemExpress

Product Code. 16418226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418226 100 mg 100mg
16402096 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16418226 Supplier Medchem Express Supplier No. HYB0294
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flubendazole is a safe and efficacious anthelmintic drug, which is widely used for anthelmintic to human, rodents and ruminants. Flubendazole exerts anticancer activities by mechanisms including inhibition of microtubule function. Flubendazole induces p53-mediated apoptosis and arrests G2/M cell cycle.

Flubendazole is a safe and efficacious anthelmintic drug, which is widely used for anthelmintic to human, rodents and ruminants. Flubendazole exerts anticancer activities by mechanisms including inhibition of microtubule function. Flubendazole induces p53-mediated apoptosis and arrests G2/M cell cycle.

Specifications

Chemical Name or Material Flubendazole
Physical Form Solid
CAS 31430-15-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H12FN3O3
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 4.17 mg/mL (13.31 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(OC)NC1=NC2=CC=C(C(C3=CC=C(F)C=C3)=O)C=C2N1
Molecular Weight (g/mol) 313.28
Formula Weight 313.28
Percent Purity 99.79%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.