missing translation for 'onlineSavingsMsg'
Learn More

Fluazifop-P-butyl, TRC

Product Code. 30398832
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398832 100 mg
30390872 1 g
30405328 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398832 Supplier Toronto Research Chemicals Supplier No. TRCF407438100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79241-46-6
Molecular Formula C19 H20 F3 N O4
Molecular Weight (g/mol) 383.36
Synonym Propanoic acid, 2-[4-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]-, butyl ester, (R)-, (+)-Fluazifop-butyl, Fluazifop-P-butyl, Fusilade 2000, Fusilade DX, Fusilade II, Fusilade Max, Fusilade S, Fusilade Super, Fusilade forte, ICI-A 0005, ICI-A 0009, IH 773B, PP 005
IUPAC Name butyl (2R)-2-[4-[5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate
SMILES CCCCOC(=O)[C@@H](C)Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H20F3NO4/c1-3-4-11-25-18(24)13(2)26-15-6-8-16(9-7-15)27-17-10-5-14(12-23-17)19(20,21)22/h5-10,12-13H,3-4,11H2,1-2H3/t13-/m1/s1
Formula Weight 383.1344
Chemical Name or Material Fluazifop-P-butyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.