Learn More
Invitrogen™ FLT4 (VEGFR3) Recombinant Human Protein
FLT4 (VEGFR3), a receptor tyrosine kinase, is a key regulator of embryonic blood vessel development and adult angiogenesis.
Brand: Invitrogen™ PR7216B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
FLT4 (VEGFR3) has been implicated in juvenile hemangioma, kaposi sarcoma, lymphangiomas, and hereditary lymphedemia.
Specifications
NP_891555 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
FLT4 (VEGFR3), PCL | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
85.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
VEGF Receptors | |
Recombinant | |
Baculovirus |