missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21049-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a class III receptor tyrosine kinase that regulates hematopoiesis. The receptor consists of an extracellular domain composed of five immunoglobulin-like domains, one transmembrane region, and a cytoplasmic kinase domain split into two parts by a kinase-insert domain. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.Specifications
| FLT3 | |
| Polyclonal | |
| Unconjugated | |
| FLT3 | |
| CD135, FL cytokine receptor, FLK2, FLT 3, FLT3, Fms like tyrosine kinase 3, fms related tyrosine kinase 3, Stem cell tyrosine kinase 1, STK 1, STK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140635, 14255, 2322 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36888, Q00342 | |
| Flt3 | |
| FLT3 Fusion Protein Ag15391 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction