Learn More
Eurofins DiscoverX Flt1 Protein, Active
Description
Two high affinity VEGF receptors, FLT1 (VEGFR-1, activated by placental growth factor PlGF) and KDR (Flk-1/VEGFR-2, activated by VEGF), have been identified. These receptors can be divided into three structural regions: seven extracellular Ig-like domains that contain the growth factor binding sites, a single polypeptide chain hydrophobic trans-membrane sequence, and intracellular cytoplasmic domains that confer the tyrosine kinase activity required for signal transduction. VEGF-B and VEGF-D homologues bind with high affinity to FLT1 (fms-related tyrosine kinase 1, vascular endothelial growth factor/vascular permeability factor receptor).
GenBank AF063657
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 89 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥58% |
| Protein | Flt1, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.