missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Flt-3/Flk-2/CD135 Antibody (IMC-EB10), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28844-1mg
This item is not returnable.
View return policy
Description
Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Flt-3/Flk-2/CD135 | |
| Monoclonal | |
| Unconjugated | |
| P36888 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| IMC-EB10 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
| FLT3/ CD135 | |
| 1 mg | |
| Cell Cycle and Replication, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase | |
| 2322 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction