Learn More
FLRT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563240
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LEIRETSFQM LPISNEPISK EEFVIHTIFP PNGMNLYKNN HSESSSNRSY RDSGIPDSDH SHS Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 53%.
This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. FLRTs may function in cell adhesion and/or receptor signaling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix. This gene is expressed in many tissues. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.Specifications
FLRT3 | |
Polyclonal | |
Unconjugated | |
FLRT3 | |
5530600M07Rik; C430047I10Rik; Fibronectin leucine rich transmembrane protein 3; fibronectin-like domain-containing leucine-rich transmembrane protein 3; FLRT3; HH21; KIAA1469; Leucine-rich repeat transmembrane protein FLRT3; mKIAA1469; RGD1566005; UNQ856/PRO1865 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23767 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZU0 | |
FLRT3 | |
Recombinant protein corresponding to Human FLRT3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only