missing translation for 'onlineSavingsMsg'
Learn More

FLRT1 Antibody (4E10), Novus Biologicals™

Product Code. 18359529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359529 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359529 Supplier Novus Biologicals Supplier No. H00023769M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FLRT1 Monoclonal antibody specifically detects FLRT1 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLRT1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4E10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH18370
Gene Alias fibronectin leucine rich transmembrane protein 1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, leucine-rich repeat transmembrane protein FLRT1, MGC21624
Host Species Mouse
Immunogen FLRT1 (AAH18370, 1 a.a. ~ 674 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVVAHPTATATTTPTATVTATVVMTTATMDLRDWLFLCYGLIAFLTEVIDSTTCPSVCRCDNGFIYCNDRGLTSIPADIPDDATTLYLQNNQINNAGIPQDLKTKVNVQVIYLYENDLDEFPINLPRSLRELHLQDNNVRTIARDSLARIPLLEKLHLDDNSVSTVSIEEDAFADSKQLKLLFLSRNHLSSIPSGLPHTLEELRLDDNRISTIPLHAFKGLNSLRRLVLDGNLLANQRIADDTFSRLQNLTELSLVRNSLAAPPLNLPSAHLQKLYLQDNAISHIPYNTLAKMRELERLDLSNNNLTTLPRGLFDDLGNLAQLLLRNNPWFCGCNLMWLRDWVKARAAVVNVRGLMCQGPEKVRGMAIKDITSEMDECFETGPQGGVANAAAKTTASNHASATTPQGSLFTLKAKRPGLRLPDSNIDYPMATGDGAKTLAIHVKALTADSIRITWKATLPASSFRLSWLRLGHSPAVGSITETLVQGDKTEYLLTALEPKSTYIICMVTMETSNAYVADETPVCAKAETADSYGPTTTLNQEQNAGPMASLPLAGIIGGAVALVFLFLVLGAICWYVHQAGELLTRERAYNRGSRKKDDYMESGTKKDNSILEIRGPGLQMLPINPYRAKEEYVVHTIFPSNGSSLCKATHTIGYGTTRGYRDGGIPDIDYSYT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23769
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.