missing translation for 'onlineSavingsMsg'
Learn More

FLRT1 Antibody (017), Alexa Fluor™ 594 , Novus Biologicals™

Product Code. 18418349 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418349 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418349 Supplier Novus Biologicals Supplier No. NBP289894AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FLRT1 Monoclonal antibody specifically detects FLRT1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLRT1
Applications ELISA
Classification Monoclonal
Clone 017
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias fibronectin leucine rich transmembrane protein 1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, leucine-rich repeat transmembrane protein FLRT1, MGC21624
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FLRT1 (Q9NZU1-1; Met1-Pro524).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23769
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.