missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ FlowX FoxP3/Transcription Factor Fixation & Perm Buffer Kit
Description
FlowX™ FoxP3/Transcription Factor Fixation & Permeabilization Buffer Kit is formulated and optimized for immunofluorescent staining of FoxP3+ or other transcription factor positive single-cell suspensions intended for flow cytometric acquisition and analysis. This product is supplemented with the metabolic inhibitor sodium azide.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Product Type | Fixative |
| Quantity | 100 Tests |
| Formulation | Supplied in in a buffered protein base with preservatives. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?