Learn More
Thermo Scientific™ Temperature Flow and Pressure Control Accessories
Shop All Thermo Scientific Products
Click to view available options
Type:
Flow Control Kit
Pressure Reducer
For Use With (Equipment):
ThermoFlex 3500 - 5000
ThermoFlex 7500 - 24000
ThermoFlex 900 - 10000
Size:
1 in. NPT
1/2 in. NPT
3/4 in. NPT
Unit Size:
Each
Description
Customize the flow and pressure output of recirculating chiller to match the needs of your process. Available in several popular sizes Thermo Scientific™ Flow and pressure control kits for Thermo Scientific chillers can have you up and running without the hassle of finding parts.
- Designed to work with your Thermo Scientific recirculating chiller
- Available in popular sizes that match your recirculating chiller connections and interface with your process
- Take the guess work out of finding parts to match your recirculating chiller connections
- Designed to match the operating range of your recirculating chiller
Additional questions? Please contact Thermo Scientific Technical support if you’re unsure how best to refine the flow and pressure of your recirculating chiller.
Specifications
Specifications
| For Use With (Equipment) | ThermoFlex 7500 - 24000 |
| Pump Type | Centrifugal Pump, Turbine Pump |
| Type | Flow Control Kit |
| Size | 1 in. NPT |