missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Flow Cytometry Staining Buffer
A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative. Designed for use in immunofluorescent staining protocols of cells in suspension.
£59.25 - £102.00
Specifications
Product Line | eBioscience™ |
---|---|
Product Type | Staining Buffer |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15346785
|
Invitrogen™
00-4222-26 |
600 mL |
£102.00
600mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15357517
|
Invitrogen™
00-4222-57 |
200 mL |
£59.25
200mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Designed for use in immunofluorescent staining protocols of cells in suspension
- A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative
- This buffer can be used for antibody and cell dilution steps, as well as all the wash steps required for the surface staining and flow cytometric analysis
- Contains animal serum proteins to help minimizing non-specific binding of antibodies
Specifications
eBioscience™ | |
Staining Buffer |
For Research Use Only. Not for use in diagnostic procedures.