missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flow Cup Viscosity Standards, Paragon Scientific™
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Manufactured in accordance with ASTM D2162 the primary method for viscosity
- Density g/mL in accordance with ASTM D1480
- Density, dynamic viscosity and kinematic viscosity quoted at all temperatures
- Ensures full compliance to ASTM & IP test method protocol
- Full Traceability to National Standards
- 2 year shelf life
Specifications
Specifications
| Viscosity | 1044 mPa.s (20°C) |
| Quantity | 500 mL |
Nominal values quoted
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?