missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flotillin 2 Antibody (PSH03-41), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32343
This item is not returnable.
View return policy
Description
Flotillin 2 Monoclonal antibody specifically detects Flotillin 2 in Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Flotillin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000-1:5000, Flow Cytometry 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Epidermal surface antigen, ESA1Membrane component chromosome 17 surface marker 1, ESAchromosome 17, surface marker 1 (35kD protein identified bymonoclonal ECS-1), flotillin 2 | |
| Recombinant protein within human Flotillin 2 aa 1-428/ 428. (Uniprot: Q14254) | |
| 100 μL | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| PSH03-41 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2319 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction