Learn More
Flotillin 2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98723APC
This item is not returnable.
View return policy
Description
Flotillin 2 Polyclonal antibody specifically detects Flotillin 2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PBS | |
| Rabbit | |
| Store at 4C in the dark. | |
| Purified |