missing translation for 'onlineSavingsMsg'
Learn More

Flotillin 2 Antibody (3G6), Novus Biologicals™

Product Code. 18392498 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392498 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392498 Supplier Novus Biologicals Supplier No. H00002319M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Flotillin 2 Monoclonal antibody specifically detects Flotillin 2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin 2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 3G6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH17292
Gene Alias Epidermal surface antigen, ESA1Membrane component chromosome 17 surface marker 1, ESAchromosome 17, surface marker 1 (35kD protein identified bymonoclonal ECS-1), flotillin 2
Host Species Mouse
Immunogen FLOT2 (AAH17292, 1 a.a. ~ 379 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTLQPRCEDVETAEGVALTVTGVAQVKIMTEKELLAVACEQFLGKNVQDIKNVVLQTLEGHLRSILGTLTVEQIYQDRDQFAKLVREVAAPDVGRMGIEILSFTIKDVYDKVDYLSSLGKTQTAVVQRDADIGVAEAERDAGIREAECKKEMLDVKFMADTKIADSKRAFELQKSAFSEEVNIKTAEAQLAYELQGAREQQKIRQEEIEIEVVQRKKQIAVEAQEILRTDKELIATVRRPAEAEAHRIQQIAEGEKVKQVLLAQAEAEKIRKIGEAEAAVIEAMGKAEAERMKLKAEAYQKYGDAAKMALVLEALPQIAAKIAAPLTKVDEIVVLSGDNSKVTSEVNRLLAELPASVHALTGVDLSKIPLIKKATGVQV
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2319
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.