missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flotillin-1 Rabbit anti-Human, Mouse, Rat, Clone: 8I2S7, Novus Biologicals™
Rabbit Monoclonal Antibody
£165.00 - £386.00
Specifications
| Antigen | Flotillin-1 |
|---|---|
| Clone | 8I2S7 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18345952
|
Bio-Techne
NBP3-16152-20UL |
20 μg |
£165.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18366506
|
Novus Biologicals
NBP3-16152-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Flotillin-1 Monoclonal antibody specifically detects Flotillin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Flotillin-1 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| flotillin 1, flotillin-1, integral membrane component of caveolae | |
| A synthetic peptide corresponding to a sequence within amino acids 328-427 of human Flotillin-1 (O75955). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8I2S7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10211 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title