Learn More
Florisil∣r, 100-200 mesh
Description
Florisil is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, anti caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Florisil is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, anti caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, hydrogen fluoride.
Specifications
Specifications
| Quantity | 1000 g |
| Mesh Size | 100 to 200 mesh |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Florisil™ |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.