missing translation for 'onlineSavingsMsg'
Learn More

Florida TRPH Standard 985 500 μg/mL in Hexane, Dr. Ehrenstorfer

Product Code. 30315030
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315030 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315030 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000985HE
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Octadecane∣n-Octane∣n-Octatriacontane∣n-Tetradecane∣n-Tetratriacontane∣Octacosane∣Tetracontane∣Tetracosane∣Triacontane
CAS 124-18-5, 629-97-0, 112-40-3, 544-85-4, 112-95-8, 630-01-3
Analyte or Component Names n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Octadecane∣n-Octane∣n-Octatriacontane∣n-Tetradecane∣n-Tetratriacontane∣Octacosane∣Tetracontane∣Tetracosane∣Triacontane
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) n-Decane 500 μg/mL∣n-Docosane 500 μg/mL∣n-Dodecane 500 μg/mL∣n-Dotriacontane 500 μg/mL∣n-Eicosane 500 μg/mL∣n-Hexacosane 500 μg/mL∣n-Hexadecane 500 μg/mL∣n-Hexatriacontane 500 μg/mL∣n-Octadecane 500 μg/mL∣n-Octane 500 μg/mL∣n-Octatriacontane 500 μg/mL∣n-Tetradecane 500 μg/mL∣n-Tetratriacontane 500 μg/mL∣Octacosane 500 μg/mL∣Tetracontane 500 μg/mL∣Tetracosane 500 μg/mL∣Triacontane 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.