missing translation for 'onlineSavingsMsg'
Learn More

Florfenicol Amine Hydrochloride, TRC

Product Code. 17960695
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960695 25 mg 25mg
17950455 10 mg 10mg
17911715 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17960695 Supplier Toronto Research Chemicals Supplier No. TRCF40577025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108656-33-3
Molecular Formula C10 H14 F N O3 S . Cl H
Molecular Weight (g/mol) 283.75
Synonym Florfenicol Amine Hydrochloride, Benzenemethanol, α-(1-amino-2-fluoroethyl)-4-(methylsulfonyl)-, hydrochloride, [R-(R*,S*)]- (9CI), Florfenicol-amine hydrochloride
IUPAC Name (1R,2S)-2-amino-3-fluoro-1-(4-methylsulfonylphenyl)propan-1-ol;hydrochloride
SMILES Cl.CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@H](N)CF

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H14FNO3S.ClH/c1-16(14,15)8-4-2-7(3-5-8)10(13)9(12)6-11;/h2-5,9-10,13H,6,12H2,1H3;1H/t9-,10-;/m1./s1
Formula Weight 283.0445
Percent Purity >95%
Chemical Name or Material Florfenicol Amine, Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.