missing translation for 'onlineSavingsMsg'
Learn More

Flopropione, MedChemExpress

Product Code. 16484376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16484376 Supplier Medchem Express Supplier No. HY100562
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flopropione is a 5-HT receptor antagonist and also a catechol-o-methyltransferase (COMT) inhibitor. Flopropione also as an antispasmodic agent.

Flopropione is a 5-HT receptor antagonist and also a catechol-o-methyltransferase (COMT) inhibitor. Flopropione also as an antispasmodic agent.

Specifications

Chemical Name or Material Flopropione
CAS 2295-58-1
Color Orange
Purity Grade Notes Research
Molecular Formula C9H10O4
Quantity 100 mg
Solubility Information DMSO : 150 mg/mL (823.41 mM; Need ultrasonic and warming)
SMILES CCC(C1=C(O)C=C(O)C=C1O)=O
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 98.93%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.