missing translation for 'onlineSavingsMsg'
Learn More

Flomoxef Sodium, TRC

Product Code. 30392239
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392239 100 mg -
30402635 500 mg -
30402259 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392239 Supplier Toronto Research Chemicals Supplier No. TRCF403000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92823-03-5
Molecular Formula C15H17F2N6NaO7S2
Molecular Weight (g/mol) 518.45
Synonym Flomoxef sodium salt, (6R-cis)-7-[[[(Difluoromethyl)thio]acetyl]amino]-3-[[[1-(2-hydroxyethyl)-1H-tetrazol-5-yl]thio]methyl]-7-methoxy-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt
SMILES [Na+].CO[C@]1(NC(=O)CSC(F)F)[C@H]2OCC(=C(N2C1=O)C(=O)[O-])CSc3nnnn3CCO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H18F2N6O7S2.Na/c1-29-15(18-8(25)6-31-13(16)17)11(28)23-9(10(26)27)7(4-30-12(15)23)5-32-14-19-20-21-22(14)2-3-24;/h12-13,24H,2-6H2,1H3,(H,18,25)(H,26,27);/q;+1/p-1/t12-,15+;/m1./s1
Formula Weight 518.0466
Chemical Name or Material Flomoxef Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.