missing translation for 'onlineSavingsMsg'
Learn More

Flometoquin, TRC

Product Code. 30398141
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398141 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398141 Supplier Toronto Research Chemicals Supplier No. TRCF11883050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flometoquin
CAS 875775-74-9
Purity Grade Notes HPLC
Molecular Formula C22 H20 F3 N O5
Quantity 50 mg
Synonym 2-Ethyl-3,7-dimethyl-6-[4-(trifluoromethoxy)phenoxy]-4-quinolinyl methyl carbonate, Flometoquin
InChI Formula InChI=1S/C22H20F3NO5/c1-5-17-13(3)20(30-21(27)28-4)16-11-19(12(2)10-18(16)26-17)29-14-6-8-15(9-7-14)31-22(23,24)25/h6-11H,5H2,1-4H3
SMILES CCc1nc2cc(C)c(Oc3ccc(OC(F)(F)F)cc3)cc2c(OC(=O)OC)c1C
IUPAC Name [2-ethyl-3,7-dimethyl-6-[4-(trifluoromethoxy)phenoxy]quinolin-4-yl] methyl carbonate
Molecular Weight (g/mol) 435.39
Formula Weight 435.1294
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.