missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flometoquin, TRC
Specifications
Specifications
| Chemical Name or Material | Flometoquin |
| CAS | 875775-74-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H20 F3 N O5 |
| Quantity | 25 mg |
| Synonym | 2-Ethyl-3,7-dimethyl-6-[4-(trifluoromethoxy)phenoxy]-4-quinolinyl methyl carbonate, Flometoquin |
| InChI Formula | InChI=1S/C22H20F3NO5/c1-5-17-13(3)20(30-21(27)28-4)16-11-19(12(2)10-18(16)26-17)29-14-6-8-15(9-7-14)31-22(23,24)25/h6-11H,5H2,1-4H3 |
| SMILES | CCc1nc2cc(C)c(Oc3ccc(OC(F)(F)F)cc3)cc2c(OC(=O)OC)c1C |
| IUPAC Name | [2-ethyl-3,7-dimethyl-6-[4-(trifluoromethoxy)phenoxy]quinolin-4-yl] methyl carbonate |
| Molecular Weight (g/mol) | 435.39 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?