Product Code. 30406431
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
30406431
1 mg
-
30390871
0.5 mg
0.5mg
30398708
5 mg
-
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
30406431
Supplier
Toronto Research Chemicals
Supplier No.
TRCF4015021MG
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Chemical Identifiers
CAS
2469039-51-6
Molecular Formula
C33H21D4F3O4
Molecular Weight (g/mol)
546.57
Synonym
4-Hydroxy-3-[1,2,3,4-tetrahydro-3-[4-[[4-(trifluoromethyl)phenyl]methoxy]phenyl]-1-naphthalenyl]-2H-1-benzopyran-2-one-d4, Storm, WL 108366-d4
SMILES
O=C1C(C(C2)C3=C(C=CC=C3)CC2C(C=C4)=CC=C4OCC5=CC=C(C(F)(F)F)C=C5)=C(O)C6=C(C([2H])=C([2H])C([2H])=C6[2H])O1
Specifications
Quantity
1 mg
InChI Formula
InChI=1S/C33H25F3O4/c34-33(35,36)24-13-9-20(10-14-24)19-39-25-15-11-21(12-16-25)23-17-22-5-1-2-6-26(22)28(18-23)30-31(37)27-7-3-4-8-29(27)40-32(30)38/h1-16,23,28,37H,17-19H2/i3D,4D,7D,8D
Percent Purity
>95%
Chemical Name or Material
Flocoumafen-d4
Grade
HPLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?