missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLNC Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-79816PE
This item is not returnable.
View return policy
Description
FLNC Polyclonal specifically detects FLNC in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| FLNC | |
| Polyclonal | |
| Flow Cytometry | |
| ABP-280, ABP280A, ABP-280-like protein, ABPA, ABP-L, ABPLABP-L, gamma filamin, Actin-binding-like protein, filamin 2, filamin C, gamma, filamin C, gamma (actin binding protein 280), Filamin-2, filamin-C, FLJ10186, FLN2actin binding protein 280, FLNc, FLN-C, Gamma-filamin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG |
| Flow Cytometry | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| FLNC | |
| Recombinant protein made to an internal portion of the human FLNC protein (amino acids 270-468) [UniProt Q14315] | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction