Learn More
FLK-1 Rat anti-Mouse, APC, Clone: Avas 12alpha1, BD
Rat Monoclonal Antibody
£325.79 - £399.65
Specifications
| Antigen | FLK-1 |
|---|---|
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Host Species | Rat |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15819908
|
BD Biosciences
560070 |
0.1 mg |
£416.00 £399.65 / Each Save £16.35 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15898048
|
BD Biosciences
555307 |
0.5 mg |
£340.00 £325.79 / Each Save £14.21 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Avas 12α1 monoclonal antibody specifically binds to fetal liver kinase 1 (Flk-1), a receptor protein tyrosine kinase closely related to CD117 (c-kit) and CD140a (PDGF Receptor α chain) of the immunoglobulin superfamily. Flk-1, also known as VEGF Receptor-2 (VEGF-R2), is a receptor for vascular endothelial growth factor (VEGF). It is expressed, at the mRNA and protein levels, on distinct sets of mesoderm during gastrulation and on endothelial cells in embryonic and adult tissues. In vivo and in vitro studies indicate that Flk-1 is required for the embryonic development of vascular endothelial and hematopoietic cells.
Specifications
| FLK-1 | |
| Monoclonal | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Primary | |
| Fetal liver kinase 1; CD309; Kdr; VEGF receptor-2; VEGFR-2 |
| Flow Cytometry | |
| Rat | |
| Mouse | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.