Learn More
Restek™ Flip Seal™ Dual Vespel™ Ring Inlet Seals
Product Code. 13129912
Encompass_Preferred
Click to view available options
Type:
Gold-Plated
Siltek-Deactivated
Packaging Quantity:
10/Pk.
2/Pk.
Product Code. 13129912
Brand: Restek™ 23413
Only null left
Add to basket
Add to basket
Reversible, two-sided design allows significantly more analyses than other seals
  • Highly inert gold or Siltek™deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column
  • Vespel™ ring embedded in top and bottom surface eliminates need for washer
  • Very little torque required to make seal, reduces operator variability
Specifications
Type Gold-Plated
Diameter (Metric) Inner 1.2 mm
For Use With (Application) Ded Design Allows Significantly More Analyses Than Other Seals
Packaging Quantity 10/Pk.