missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLIP Antibody (NF6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-80081IR
This item is not returnable.
View return policy
Description
FLIP Monoclonal antibody specifically detects FLIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| FLIP | |
| Monoclonal | |
| DyLight 755 | |
| CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta | |
| Recombinant human FLIP (aa 1-480). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Inhibitors Activators and Regulators, Signal Transduction, Tumor Suppressors | |
| 8837 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| NF6 | |
| 50mM Sodium Borate | |
| Mouse | |
| >95%, by SDS-PAGE | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction