missing translation for 'onlineSavingsMsg'
Learn More

FLIP Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18419502 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419502 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419502 Supplier Novus Biologicals Supplier No. NBP177016JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FLIP Polyclonal antibody specifically detects FLIP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLIP
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the amino terminus of human FLIP. The sequence is identical in all FLIP splice variants. The immunogen is located within the first 50 amino acids of FLIP. Amino Acid Squence: SAEVIHQVEEALDTDEK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Inhibitors Activators and Regulators, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8837
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.