missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLII Recombinant Rabbit Monoclonal Antibody (K01_3K84)
Description
FLII Recombinant Monoclonal Antibody for Western Blot
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
Specifications
Specifications
| Antigen | FLII |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_3K84 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | FLII |
| Gene Accession No. | Q13045, Q9JJ28 |
| Gene Alias | 3632430F08Rik; FLI; Fli1; flightless I actin binding protein; flightless I homolog; flightless I homolog (Drosophila); Flii; FLII actin remodeling protein; FLII, actin remodeling protein; Fliih; FLIL; Protein flightless-1 homolog |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?