missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLII Recombinant Rabbit Monoclonal Antibody (HL2966)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556711
This item is not returnable.
View return policy
Description
FLII Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
Specifications
| FLII | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q13045, Q9JJ28 | |
| FLII | |
| Recombinant fragment of human Flightless I. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2966 | |
| Unconjugated | |
| FLII | |
| 3632430F08Rik; FLI; Fli1; flightless I actin binding protein; flightless I homolog; flightless I homolog (Drosophila); Flii; FLII actin remodeling protein; FLII, actin remodeling protein; Fliih; FLIL; Protein flightless-1 homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14248, 2314, 287375 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction