missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLII Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521735
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, H1299, HeLaS3, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (81%), Chicken (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
Specifications
| FLII | |
| Polyclonal | |
| Unconjugated | |
| FLII | |
| 3632430F08Rik; FLI; Fli1; flightless I actin binding protein; flightless I homolog; flightless I homolog (Drosophila); Flii; FLII actin remodeling protein; FLII, actin remodeling protein; Fliih; FLIL; Protein flightless-1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2314 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.88 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q13045 | |
| FLII | |
| Recombinant protein encompassing a sequence within the center region of human Flightless I. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction