Learn More
Flightless I Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27729-1-AP-20UL
Description
The Flightless I antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Flightless I. This antibody recognizes human antigen. The Flightless I antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Flightless I | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| FLI, Fli1, Flightless-1, FLII, FLIL, Protein flightless 1 homolog | |
| 20 μL | |
| 2314 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |