missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Mouse anti-Human, Clone-FLI1/1312, Novus Biologicals™
Mouse Monoclonal Antibody
£529.00
Specifications
| Antigen | FLI1 |
|---|---|
| Clone | FLI1/1312 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF |
Description
FLI1 Monoclonal specifically detects FLI1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| FLI1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
| FLI1 | |
| IgG | |
| Protein A purified | |
| 51 kDa |
| FLI1/1312 | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cancer, Tumor Suppressors | |
| PBS with No Preservative | |
| 2313 | |
| Recombinant fragment (aa 107-261) of human FLI1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title