missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLG Mouse anti-Human, Clone: FLG/1561, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14526.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against partial recombinant human FLG.
- High specificity
- High reactivity
Specifications
| FLG | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P20930 | |
| FLG | |
| Recombinant protein corresponding to amino acids 998-1104 of human FLG. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| FLG/1561 | |
| Mouse monoclonal antibody raised against partial recombinant human FLG. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| ATOD2 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 2312 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction