missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Flexsafe 2D Bags, 50 mL

Product Code. 15914570 Shop All Sartorius Products
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15914570 50 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15914570 Supplier Sartorius Stedim Biotech™ Supplier No. FLS130114

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Flexible sampling bag designed for sanitary or aseptic sampling.

  • Flexible bag with dual filling/draining lines
  • Large sealing window for robustness
  • Puncture resistant
  • Excellent biocompatibility
  • Inert PE contact layer
  • High gas and water barrier properties
  • Low extractable profile
  • Suitable for use with the following:
  • Media
  • Buffer
  • Cell harvest and DSP
  • Drug substances
  • Drug products
TRUSTED_SUSTAINABILITY

Specifications

Includes Inlet: 1/4 in. MPC female plus sealing cap, Outlet: 1/4 in. MPC male plus sealing cap
Material Polyethylene
Tubing Style Silicone tubing, 150 mm
Capacity (Metric) 50 mL
Thickness (Metric) 400 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.