Learn More
Honeywell™ FLEXIDYN Light Task Plus 4 Gloves
Description
- Nylon and glass fiber knitted glove 3/4 Bi polymer (PU and Nitrile) coating knitted wrist
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The Glass fiber protected by polyamide thread provides very good properties against cuts
- The 3/4 Bi polymer coating provides outstanding abrasion and perforation resistance and protects well the knuckle
- Its Dark color enables work in a slightly soiled environment
Specifications
Specifications
| Certifications/Compliance | EC PPE Category 2, EC Certificate Number 0075/797/162/06/18/1075 |
| Description | 10 gauge |
| Color | Gray, Blue |
| Material | Nylon, Glass Fiber |
| Size | 7, Small |
| Length (Metric) | 25 cm |