Learn More
Sartorius Flexible Sample Holder, for Cubis™ Balances
For use with Cubis Balances. Q-Grip is a flexible and adaptable “one-size-fits-all” holder for bottles, test tubes, reaction containers or filters (up to 120mm) for all Cubis semi-microbalances and analytical balances.
Brand: Sartorius YFH01MS
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Includes:
- Plate
- Filter supports (2)
- Flask supports (2)
- Pan support
- Clamping screw and nut
Replaces the original weighing pan and is only for CUBIS Balances with 0.1 and 0.01mg readability.
Specifications
Containers and Filters of Cubis Semi-microbalance | |
Weighting Flask Holder |