missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UN75plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN75plus Oven
Unit Size:
Each
Description
- Positioned flexibly in chamber or load
- For local temperature measurement (up to 1 additional sensor is possible)
- The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
- For Memmert Ovens
Specifications
Specifications
| For Use With (Equipment) | UN75plus Oven |
| Type | Temperature Sensor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction