missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Flexible Pt100 Temperature Sensor for UN30

Product Code. 17540096 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17540096

Brand: Memmert™ UN30H8

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Flexible temperature sensor for positioning in chamber or in load.

  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens

Specifications

For Use With (Equipment) UN30 Oven
Type Temperature Sensor
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.