Learn More
Memmert™ Flexible Pt100 For Chamber Or Load, Display And Atmocontrol Compatible, For Un110Pa
Shop All Memmert ProductsDescription
Includes
4-Pin Connector for Temperature Recording
- Can be positioned in chamber or in load
- Includes 4-pin connector, according to NAMUR NE 28, for external temperature recording (load temperature) max. 3 sensors
- For use in Memmert™ universal ovens
Specifications
Specifications
| Description | Secondary/Additional PT 100 Sensor (up to 3 can be attached in total) |
| For Use With (Equipment) | Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108 |
| Type | Temperature Sensor |
| For Use With (Application) | Load/Chamber Temperature Measurement |