missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Flexible Duckboard

Product Code. 30245722
Click to view available options
:
Gilac™ Flexible Duckboard
This item is not returnable. View return policy

Product Code. 30245722

Brand: Gilac™ G402222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flexible duckboard in low-density polyethylene. Open structure for optimum drainage of liquids.

  • Essential for identifying storage areas by color. Allows compliance with hygiene rules for food storage
  • High load capacity thanks to HDPE structure
  • Easy to clean with a high pressure water jet or in the machine
  • Clip -in system very easy to install, easily cut with a saw for a perfect fit in its slot
  • Junction area suitable for wide surfaces equipment
  • Modular, upgradeable and durable
  • Thermal resistance suitable for temperatures in freezing chambers
  • Optimum drainage

Specifications

Closure Type None
Color Gray
Material HDPE
Shape Square
Product Type Flexible Duckboard
Dimensions (L x W x H) 500 x 500 x 50 mm
Length (Metric) 500 mm
Width (Metric) 500 mm
Height (Metric) 50 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.