missing translation for 'onlineSavingsMsg'
Learn More

Leica Microsystems Flexacam i5

Product Code. 17873438
Change view
Click to view available options
For Use With (Equipment):
Compound Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17873438 Compound Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17873438 Supplier Leica Microsystems Supplier No. 12730537
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color camera for compound microscopes with high definition live image

  • Digital color camera with CMOS sensor (1/2.3) in.
  • Possible to operate with 4k HDMI monitor using Enersight OSD
  • High definition live image 4k, 60 fps, HDR
  • JPG images with up to 12 MP
  • Full HD MJPG Video recording with 30 fps
  • Camera control: via PC (Windows) using Enersight Desktop, via smartphone / tablet by Enersight Mobile App (connection to Ethernet or Wifi is required) or via USB mouse (not included) (Enersight OSD mode)
  • Capture images directly to USB Stick or send images via E-Mail (Enersight OSD mode)
  • USB-C connection to PC for Enersight Desktop
  • Supported OS: Win 10 Pro/ Win 11 / iOS / Android
  • Delivered accessories: HDMI Cable, USB C-A cable, USB stick
TRUSTED_SUSTAINABILITY

Specifications

Type Camera
For Use With (Equipment) Compound Microscopes
Includes HDMI Cable, USB C-A cable, USB stick
Port Type USB C-A
Sensor Types CMOS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.