missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fleroxacin, MedChemExpress
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Description
Fleroxacin (RO 23-6240) is a broad-spectrum antimicrobial fluoroquinolone.
Specifications
Specifications
| Chemical Name or Material | Fleroxacin |
| Physical Form | Solid |
| CAS | 79660-72-3 |
| Color | White |
| Quantity | 10 g |
| Purity Grade Notes | Research |
| Molecular Formula | C17H18F3N3O3 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | RO 23-6240 AM-833 |
| Solubility Information | 0.1 M NaOH : 9.17 mg/mL (24.83 mM; ultrasonic and adjust pH to 12 with NaOH) ∣H2O : < 0.1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction