missing translation for 'onlineSavingsMsg'
Learn More

Flecainide Acetate, 98%, Thermo Scientific™

Sodium channel blocker

£164.00 - £453.01

Chemical Identifiers

CAS 54143-56-5
Molecular Formula C19H24F6N2O5
Molecular Weight (g/mol) 474.4
MDL Number MFCD00214290
InChI Key RKXNZRPQSOPPRN-UHFFFAOYSA-N
Synonym flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
PubChem CID 41022
ChEBI CHEBI:5091
IUPAC Name acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
SMILES CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15445739
SDS View Documents
Thermo Scientific Alfa Aesar
J63527.MC
100mg
ONLINE EXCLUSIVE PRICE
£168.00
£164.00 / 100mg
Save £4.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15435739
SDS View Documents
Thermo Scientific Alfa Aesar
J63527.03
1g
ONLINE EXCLUSIVE PRICE
£476.00
£453.01 / 1g
Save £22.99
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in dilute acetic acid, dimethyl sulfoxide, ethanol, methanol and water.

Notes
Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

54143-56-5
474.4
RKXNZRPQSOPPRN-UHFFFAOYSA-N
41022
acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
C19H24F6N2O5
MFCD00214290
flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
CHEBI:5091
CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F

Specifications

54143-56-5
MFCD00214290
flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
RKXNZRPQSOPPRN-UHFFFAOYSA-N
acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
41022
474.4
Flecainide acetate
C19H24F6N2O5
14,4098
Soluble in dilute acetic acid,dimethyl sulfoxide,ethanol,methanol and water.
CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F
474.4
CHEBI:5091
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H361

missing translation for 'einecsNumber' : 258-997-5

missing translation for 'rtecsNumber' : CV5792550

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.